Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenyliodonium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

A17934-30 A17934-18 A17934-30 A17934-09 A17934-18 A17934-09
AAAA17934-30EA 1334.99 USD InStock
CAAAAA17934-18 CAAAAA17934-30 AAAA17934-30 AAAA17934-18 CAAAAA17934-09 AAAA17934-09
Diphenyliodonium hexafluorophosphate 98%
Diphenyliodonium hexafluorophosphate


Formula: C₁₂H₁₀F₆IP
Melting Pt: 139…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00061398
CAS Number: 58109-40-3
EINECS: 261-134-5
UN: 1759
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty