Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2-Diphenylpropiononitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

A17991-18 A17991-09 A17991-06 A17991-09 A17991-06 A17991-18
AAAA17991-06EA 31.09 USD InStock
CAAAA17991-09 CAAAA17991-18 CAAAAA17991-06 AAAA17991-06 AAA17991-09 AAA17991-18
2,2-Diphenylpropiononitrile 97%
2,2-Diphenylpropiononitrile


Formula: (C₆H₅)₂C(CN)CH₃
MW: 207.27 g/mol
Boiling Pt: 144…145 °C/2 mm
Density: 1.108
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001846
CAS Number: 5558-67-8
EINECS: 226-925-1
UN: 2810
ADR: "6.1",III
https://us-legacy.vwr.com/stibo/web/std.lang.all/68/42/2296842.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty