Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenylbutyric acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Phenylbutanoic acid

A18115-36 A18115-36 A18115-22 A18115-14 A18115-14 A18115-22
AAAA18115-36EA 850.99 USD InStock
CAAAAA18115-36 AAAA18115-36 AAA18115-14 AAA18115-22 CAAAA18115-14 CAAAA18115-22
4-Phenylbutyric acid 99%
4-Phenylbutyric acid


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 165 °C/10 mm
Melting Pt: 49…53 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty