Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-PhenylbenzeneamineN-Phenylaniline

A18265-36 A18265-22 A18265-0E
AAAA18265-0EEA 193.99 USD InStock
AAAA18265-0E AAAA18265-36 AAAA18265-22
Diphenylamine 98+%
Diphenylamine


Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302 °C
Melting Pt: 52…55 °C
Density: 1.16
Flash Pt: 152 °C(305 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: "9",III
Merck Index: 13,03349
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/36/2297636.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty