Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium-2-ethylhexanoate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Ethylhexanoic acid calcium saltCalcium 2-ethylhexanoate2-Ethylcaproic acid calcium salt

A18266-14 A18266-06 A18266-22 A18266-22 A18266-14 A18266-06
AAAA18266-06EA 56.59 USD InStock
AAAA18266-06 AAAA18266-14 CAAAAA18266-06 CAAAAA18266-22 CAAAAA18266-14 AAAA18266-22
Calcium-2-ethylhexanoate 98%
Calcium 2-ethylhexanoate


Formula: C₁₆H₃₀CaO₄
MW: 326.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014001
CAS Number: 136-51-6
EINECS: 205-249-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty