Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hydrogenarsenate heptahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Arsenic Acid, Sodium Salt Heptahydrate. Sodium SaltSodium arsenate dibasic heptahydrateSodium hydrogen arsenate heptahydrate

A18275-36 A18275-22 A18275-36 A18275-22
AAAA18275-36EA 288.99 USD InStock
CAAAAA18275-22 AAAA18275-36 AAAA18275-22 CAAAAA18275-36
Disodium hydrogenarsenate heptahydrate 98%
Disodium hydrogenarsenate heptahydrate


Formula: HAsNa₂O₄·7H₂O
MW: 312.01 g/mol
Melting Pt: 130 °C (dec.)
Density: 1.88
Storage Temperature: Ambient
MDL Number: MFCD00149179
CAS Number: 10048-95-0
EINECS: 231-902-4
UN: 1685
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty