Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pimelic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,5-Pentanedicarboxylic acidHeptanedioic acid1,7-Heptanedioic acid

A18495-14 A18495-22 A18495-22 A18495-14
AAAA18495-22EA 133.92 USD InStock
CAAAAA18495-22 CAAAAA18495-14 AAAA18495-22 AAAA18495-14
Pimelic acid 98+%
Pimelic acid


Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 212 °C/10 mm
Melting Pt: 102…106 °C
Density: 1,329
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0
EINECS: 203-840-8
Merck Index: 12,07585
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty