Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Acetyl-2-naphthol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(2-Hydroxy-1-naphthyl)ethan-1-one 2-Hydroxy-1-acetonaphthone 1-Acetyl-2-hydroxynaphthalene

A18537-14 A18537-06 A18537-22 A18537-22 A18537-06 A18537-14
AAAA18537-06EA 26.49 USD InStock
CAAAA18537-22 AAAA18537-06 AAA18537-14 AAA18537-22 CAAAAA18537-06 CAAAA18537-14
1-Acetyl-2-naphthol 99%
1-Acetyl-2-naphthol


Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 130…132 °C/2 mm
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00012131
CAS Number: 574-19-6
EINECS: 209-369-4
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty