Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,6,6-Tetramethylpiperidine 98+%

Supplier: Thermo Scientific Chemicals
Danger

A18712-22 A18712-22 A18712-06 A18712-14 A18712-14 A18712-06
AAAA18712-22EA 428.99 USD InStock
AAAA18712-22 AAA18712-06 AAA18712-14 CAAAA18712-06 CAAAA18712-14 CAAAAA18712-22
2,2,6,6-Tetramethylpiperidine 98+%
2,2,6,6-Tetramethylpiperidine


Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 152…154 °C
Melting Pt: –59 °C
Density: 0.832
Flash Pt: 34 °C(93 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005985
CAS Number: 768-66-1
EINECS: 212-199-3
UN: 1992
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty