Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amidosulfonic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Aminosulphonic acidAmidosulphonic acidSulphamic acidAmidosulfuric AcidAmidosulphuric AcidSulphamidic acid

A18836-36 A18836-0E A18836-22 A18836-22 A18836-0E A18836-36
AAAA18836-22EA 48.76 USD InStock
CAAAAA18836-22 AAAA18836-22 CAAAAA18836-36 CAAAAA18836-0E AAAA18836-36 AAAA18836-0E
Amidosulfonic acid 98+%
Amidosulphonic acid


Formula: H₃NO₃S
MW: 97.09 g/mol
Melting Pt: ca 205 °C (dec.)
Density: 2.12
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: "8",III
Merck Index: 13,09006
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/87/2297587.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty