Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-fluoro-4-iodobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

A19190-14 A19190-06 A19190-22 A19190-22 A19190-06 A19190-14
AAA19190-14EA 244.99 USD InStock
CAAAA19190-14 CAAAA19190-06 AAA19190-14 AAAA19190-22 CAAAAA19190-22 AAA19190-06
1-Bromo-2-fluoro-4-iodobenzene 97%
1-Bromo-2-fluoro-4-iodobenzene


Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 80 °C/2.5 mm
Melting Pt: 34…38 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042577
CAS Number: 136434-77-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty