Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Diethoxy-3-cyclobutene-1,2-dione 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Diethyl squarate

A19248-03 A19248-06 A19248-14 A19248-03 A19248-14 A19248-06
AAAA19248-14EA 698.99 USD InStock
CAAAA19248-03 CAAAAA19248-14 AAAA19248-14 CAAAA19248-06 AAA19248-06 AAA19248-03
3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
3,4-Diethoxy-3-cyclobutene-1,2-dione


Formula: C₈H₁₀O₄
MW: 170.16 g/mol
Boiling Pt: 95 °C/0.1 mm
Density: 1.162
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001333
CAS Number: 5231-87-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty