Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diisopropylcarbodiimide 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N,N'-methanetetraylbis(1-methylethylamine) N,N'-Diisopropylcarbodiimide 1,3-Diisopropylcarbodiimide DIC

A19292-09 A19292-30 A19292-14 A19292-30 A19292-18 A19292-14 A19292-09 A19292-18
AAAA19292-30EA 597.99 USD InStock
CAAAAA19292-18 CAAAA19292-14 CAAAAA19292-30 AAAA19292-30 AAAA19292-18 CAAAAA19292-09 AAAA19292-09 AAA19292-14
Diisopropylcarbodiimide 99%
Diisopropylcarbodiimide


Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 145…148 °C
Density: 0.810
Flash Pt: 33 °C(91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00065689
CAS Number: 693-13-0
EINECS: 211-743-7
UN: 3384
ADR: "6.1",I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty