Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isovaleraldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Methylbutyraldehyde

A19640-AU A19640-AP A19640-AU A19640-AE A19640-AE A19640-AP
AAA19640-AUEA 161.99 USD InStock
AAA19640-AU CAAAAA19640-AP CAAAA19640-AU AAAA19640-AP CAAAAA19640-AE AAAA19640-AE
Isovaleraldehyde 98%
Isovaleraldehyde


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 91…93 °C
Melting Pt: –51 °C
Density: 0.800
Flash Pt: -1 °C(30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007014
CAS Number: 590-86-3
EINECS: 209-691-5
UN: 2058
ADR: "3",II
Merck Index: 13,05250
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty