Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenanthrene tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

A19646-36 A19646-36 A19646-0E A19646-22 A19646-22 A19646-0E
AAAA19646-22EA 71.99 USD InStock
CAAAAA19646-22 CAAAAA19646-36 AAAA19646-22 AAAA19646-0E AAAA19646-36 CAAAAA19646-0E
Phenanthrene tech. 90%, Technical Grade
Phenanthrene


Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340 °C
Melting Pt: 97…101 °C
Density: 0.980
Flash Pt: 171 °C(339 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001168
CAS Number: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: "9",III
Merck Index: 12,07354
https://us-legacy.vwr.com/stibo/web/std.lang.all/84/56/2298456.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty