Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Hydroxyquinoline-5-sulfonic acid hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

B20022-18 B20022-36 B20022-36 B20022-30 B20022-18 B20022-30
AAAB20022-36EA 175.99 USD InStock
CAAAAB20022-36 CAAAB20022-18 AAAB20022-36 AAB20022-18 AAB20022-30 CAAAB20022-30
8-Hydroxyquinoline-5-sulfonic acid hydrate 98%
8-Hydroxyquinoline-5-sulphonic acid hydrate


Formula: C₉H₇NO₄S·H₂O
MW: 225.22 (anhy)
Melting Pt: 311…313 °C
Storage Temperature: Ambient
MDL Number: MFCD00149482
CAS Number: 207386-92-3
EINECS: 201-570-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty