Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-fluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-4-fluoro-1-methylbenzene

B20278-36 B20278-22 B20278-14 B20278-14 B20278-22 B20278-36
AAB20278-14EA 30.19 USD InStock
CAAAB20278-14 CAAAB20278-22 AAB20278-14 CAAAAB20278-36 AAAB20278-36 AAB20278-22
2-Chloro-4-fluorotoluene 98%
2-Chloro-4-fluorotoluene


Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 154…156 °C
Density: 1.198
Flash Pt: 50 °C(122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000572
CAS Number: 452-73-3
EINECS: 207-209-8
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty