Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palmitic acid 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Hexadecanoic acidHexadecylic acid

B20322-0C B20322-36 B20322-36 B20322-0C B20322-0E B20322-0E
AAAB20322-0EEA 111.56 USD InStock
CAAAAB20322-0E AAAB20322-0E CAAAAB20322-36 CAAAAB20322-0C AAAB20322-0C AAAB20322-36
Palmitic acid 95%
Palmitic acid
A saturated fatty acid


Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 215 °C/15 mm
Melting Pt: 62…65 °C
Density: 0.853
Flash Pt: 206 °C(402 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty