Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Iodoperfluorohexane 98+% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-6-iodohexane Perfluorohexyl iodide

Stabilization: Stabilised with copper

B20958-18 B20958-18 B20958-30 B20958-30 B20958-09 B20958-09
AAB20958-09EA 45.59 USD InStock
AAB20958-09 AAB20958-18 AAAB20958-30 CAAAB20958-18 CAAAB20958-09 CAAAAB20958-30
1-Iodoperfluorohexane 98+% stabilized
1-Iodoperfluorohexane


Formula: C₆F₁₃I
MW: 445.95 g/mol
Boiling Pt: 117 °C
Melting Pt: –45 °C
Density: 2.063
Storage Temperature: Ambient
MDL Number: MFCD00001063
CAS Number: 355-43-1
EINECS: 206-586-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty