Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(diethylamino)chlorophosphine 94%

Supplier: Thermo Scientific Chemicals
Danger

B21019-06 B21019-22 B21019-22 B21019-14 B21019-14 B21019-06
AAAB21019-14EA 673.99 USD InStock
AAAB21019-14 CAAAAB21019-06 AAAB21019-06 CAAAAB21019-22 AAAB21019-22 CAAAAB21019-14
Bis(diethylamino)chlorophosphine 94%
Bis(diethylamino)chlorophosphine


Formula: C₈H₂₀ClN₂P
Boiling Pt: 55…57 °C/0.2 mm
Density: 1.002
Flash Pt: 101 °C(214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015493
CAS Number: 685-83-6
UN: 1760
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty