Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Histidine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R)-(-)-Histidine (R)-(-)-Histidine D-3-Imidazol-4-ylalanine H-D-His-OH

B21027-14 B21027-22 B21027-22 B21027-06 B21027-06 B21027-14
AAB21027-06EA 100.02 USD InStock
CAAAAB21027-22 CAAAB21027-06 AAB21027-06 AAB21027-14 AAAB21027-22 CAAAB21027-14
D-(-)-Histidine 99%
D(-)-Histidine


Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: ca 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00065963
CAS Number: 351-50-8
EINECS: 206-513-8
Merck Index: 12,04758
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty