Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Serotonin hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Hydroxytryptamine Hydrochloride

B21263-09 B21263-03 B21263-06 B21263-03 B21263-09 B21263-06
AAAB21263-09EA 868.99 USD InStock
AAAB21263-09 CAAAAB21263-09 CAAAB21263-03 CAAAB21263-06 AAB21263-03 AAB21263-06
Serotonin hydrochloride 98%
Serotonin hydrochloride


Formula: C₁₀H₁₃ClN₂O
MW: 212.68 g/mol
Melting Pt: 167…172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012686
CAS Number: 153-98-0
EINECS: 244-464-4
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty