Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methylcyclohexanecarboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Methylcyclohexane-1-carboxylic acid

B21713-14 B21713-06 B21713-14 B21713-06 B21713-18 B21713-18
AAB21713-14EA 161.99 USD InStock
CAAAAB21713-18 AAB21713-14 AAB21713-06 CAAAB21713-14 AAAB21713-18 CAAAB21713-06
1-Methylcyclohexanecarboxylic acid 99%
1-Methylcyclohexanecarboxylic acid


Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 236 °C
Melting Pt: 36…39 °C
Density: 1.0115 g/cm³ (20 °C)
Flash Pt: 100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001463
CAS Number: 1123-25-7
EINECS: 214-371-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty