Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-DL-methionine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-DL-Met-OH

B21866-18 B21866-18 B21866-0B B21866-30 B21866-0B B21866-30
AAB21866-0BEA 97.09 USD InStock
AAB21866-0B CAAAB21866-0B AAAB21866-18 AAB21866-30 CAAAB21866-30 CAAAAB21866-18
N-Acetyl-DL-methionine 99%
N-Acetyl-DL-methionine


Formula: C₇H₁₃NO₃S
MW: 191.2 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008681
CAS Number: 1115-47-5
EINECS: 214-224-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty