Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) sulfate tetrahydrate 99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Manganese sulphate tetrahydrateManganous sulfate tetrahydrateManganese sulfate tetrahydrateManganous sulphate tetrahydrate

B22081-36 B22081-22 B22081-36 B22081-22
AAAB22081-36EA 126.8 USD InStock
AAAB22081-36 CAAAAB22081-36 AAAB22081-22 CAAAAB22081-22
Manganese(II) sulfate tetrahydrate 99% (metals basis)
Manganese (II) sulphate tetrahydrate


Formula: MnO₄S·4H₂O
MW: 223.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149160
CAS Number: 10101-68-5
EINECS: 232-089-9
UN: 3077
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty