Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Phenylglycine ethyl ester 99%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Phenylglycine Ethyl EsterPhenylamino-acetic acid ethyl ester

B22282-30 B22282-09 B22282-18 B22282-09 B22282-30 B22282-18
AAAB22282-30EA 894.9 USD InStock
CAAAAB22282-09 CAAAAB22282-30 AAAB22282-30 AAAB22282-09 AAAB22282-18 CAAAAB22282-18
N-Phenylglycine ethyl ester 99%
N-Phenylglycine ethyl ester


Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: 55…58 °C
Storage Temperature: Refrigerator
EINECS: 218-702-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty