Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Mercaptomenthone cis- and trans- mixture 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 8-Mercaptomenthonep-Mentha-8-thiol-3-one2-(1-Mercapto-1-methylethyl)-5-methylcyclohexanone

B22287-14 B22287-14 B22287-06 B22287-06
AAAB22287-06EA 64.59 USD InStock
CAAAAB22287-14 AAAB22287-06 AAAB22287-14 CAAAAB22287-06
8-Mercaptomenthone cis- and trans- mixture 97%
8-Mercaptomenthone


Formula: C₁₀H₁₈OS
MW: 186.31 g/mol
Boiling Pt: 270…272 °C
Density: 1.00
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012393
CAS Number: 38462-22-5
EINECS: 253-953-1
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty