Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloridep-Iodonitrotetrazolium violetIodonitrotetrazolium violet3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyltetrazolium chloride2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chlorideIodonitrotetrazolium chlorideINT

B22301-03 B22301-14 B22301-06 B22301-14 B22301-03 B22301-06
AAAB22301-14EA 858.99 USD InStock
CAAAAB22301-03 CAAAAB22301-14 CAAAAB22301-06 AAAB22301-14 AAAB22301-03 AAAB22301-06
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride 95%
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Used for dehydrogenase determination


Formula: C₁₉H₁₃ClIN₅O₂
MW: 505.7 g/mol
Melting Pt: ca 231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011961
CAS Number: 146-68-9
EINECS: 205-676-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty