Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bathocuproin sulfonate disodium salt hydrate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Bathocuproindisulphonic disodium salt hydrate

B22550-06 B22550-03 B22550-06 B22550-03 B22550-MD B22550-MD
AAAB22550-03EA 118.14 USD InStock
CAAAAB22550-03 CAAAAB22550-MD CAAAAB22550-06 AAAB22550-03 AAAB22550-MD AAAB22550-06
Bathocuproin sulfonate disodium salt hydrate 97%
Bathocuproin sulfonate disodium salt hydrate
Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated procedures


Formula: C₂₆H₁₈N₂Na₂O₆S₂·H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03012947
CAS Number: 52698-84-7
EINECS: 258-111-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty