Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

B22773-22 B22773-30 B22773-22 B22773-30
AAAB22773-22EA 67.09 USD InStock
CAAAB22773-30 AAAB22773-22 CAAAAB22773-22 AAB22773-30
1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
1-Chloro-3,4-dinitrobenzene


Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Boiling Pt: 14…16 °C/4 mm
Melting Pt: ca 20…25 °C
Density: 1.687
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007212
CAS Number: 610-40-2
EINECS: 210-223-7
UN: 1577
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty