Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Bromopropan-1-ol1-Bromo-3-hydroxypropane

B22857-22 B22857-06 B22857-22 B22857-14 B22857-14 B22857-06
AAAB22857-06EA 35.69 USD InStock
CAAAAB22857-06 AAAB22857-06 AAAB22857-22 CAAAAB22857-14 AAAB22857-14 CAAAAB22857-22
3-Bromo-1-propanol 97%
3-Bromo-1-propanol


Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 71…73 °C (11 mmHg)
Density: 1,590
Flash Pt: 93 °C (199 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty