Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2R)-(+)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

B22968-ME B22968-ME B22968-77 B22968-77
AAB22968-77EA 51.89 USD InStock
AAB22968-77 CAAAAB22968-ME AAAB22968-ME CAAAB22968-77
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid


Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 105…107 °C/1 mm
Melting Pt: 43…45 °C
Density: 1,344
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004184
CAS Number: 20445-31-2
EINECS: 243-829-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty