Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloromethylvinylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Vinylmethyldichlorosilane

B23119-36 B23119-36
AAAB23119-36EA 119.99 USD InStock
AAAB23119-36 CAAAAB23119-36
Dichloromethylvinylsilane 97%
Dichloromethylvinylsilane


Formula: C₃H₆Cl₂Si
Boiling Pt: 92…93 °C
Melting Pt: –78 °C
Density: 1.087
Flash Pt: 4 °C(39 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000492
CAS Number: 124-70-9
EINECS: 204-710-3
UN: 2985
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty