Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Aminoethylamino)propylmethyldimethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-(2-Aminoethylamino)propyldimethoxymethylsilane

B23159-22 B23159-36 B23159-14 B23159-36 B23159-22 B23159-14
AAAB23159-22EA 66.99 USD InStock
AAAB23159-22 CAAAAB23159-14 CAAAAB23159-36 CAAAAB23159-22 AAAB23159-36 AAAB23159-14
3-(2-Aminoethylamino)propylmethyldimethoxysilane 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane


Formula: C₈H₂₂N₂O₂Si
MW: 206.36 g/mol
Boiling Pt: 265 °C
Density: 0.968
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054776
CAS Number: 3069-29-2
EINECS: 221-336-6
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty