Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate4,7-Diphenyl-1,10-phenanthrolinedisulphonic acid disodium salt hydrate

B23244-MD B23244-06 B23244-MD B23244-06 B23244-03 B23244-03
AAAB23244-MDEA 49.19 USD InStock
AAAB23244-MD CAAAAB23244-MD AAAB23244-06 CAAAAB23244-06 AAAB23244-03 CAAAAB23244-03
Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%
Bathophenanthrolinedisulphonic acid disodium salt hydrate


Formula: C₂₄H₁₄N₂Na₂O₆S₂·H₂O
MW: 536.48 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149310
CAS Number: 52746-49-3
EINECS: 258-152-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty