Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Patton-Reeders reagent (Calconcarboxylic acid), (max. 10% H₂O)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid3-Hydroxy-4-(2-hydroxy-4-sulpho-1-naphthylazo)naphthalene-2-carboxylic acid2-Hydroxy-1-(2-hydroxy-4-sulpho-1-naphthylazo)-3-naphthoic acid2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acidCal-Red®3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalene-2-carboxylic acid2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalene-3-carboxylic acid

B23459-09 B23459-09 B23459-18 B23459-18
AAAB23459-09EA 79.53 USD InStock
AAAB23459-09 CAAAAB23459-18 AAAB23459-18 CAAAAB23459-09
Patton-Reeders reagent (Calconcarboxylic acid), (max. 10% H₂O)
Patton-reeders reagent (Calconcarboxylic acid)


Formula: C₂₁H₁₄N₂O₇S
MW: 438.42 g/mol
Melting Pt: ca 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004078
CAS Number: 3737-95-9
EINECS: 223-117-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty