Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Cyano-2-nitroaniline

B23463-03 B23463-03 B23463-14 B23463-14 B23463-06 B23463-06
AAAB23463-06EA 137.99 USD InStock
CAAAAB23463-03 CAAAAB23463-14 AAAB23463-06 AAAB23463-03 AAAB23463-14 CAAAAB23463-06
4-Amino-3-nitrobenzonitrile 98%
4-Amino-3-nitrobenzonitrile


Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00013373
CAS Number: 6393-40-4
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty