Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Bis(trifluoromethyl)styrene 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

B23499-03 B23499-06 B23499-03 B23499-06
AAAB23499-03EA 113.85 USD InStock
CAAAAB23499-03 CAAAAB23499-06 AAAB23499-03 AAAB23499-06
3,5-Bis(trifluoromethyl)styrene 98% stabilized
3,5-Bis(trifluoromethyl)styrene


Formula: C₁₀H₆F₆
MW: 240.15 g/mol
Boiling Pt: 59…61 °C/20 mm
Melting Pt: 7…8 °C
Density: 1.334
Flash Pt: 50 °C(122 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00075538
CAS Number: 349-59-7
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty