Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (3-Fluorophenyl)methanamine

B23527-14 B23527-14 B23527-06 B23527-06
AAAB23527-06EA 72.29 USD InStock
AAAB23527-06 AAAB23527-14 CAAAAB23527-06 CAAAAB23527-14
3-Fluorobenzylamine 97%
3-Fluorobenzylamine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 186 °C
Density: 1.097
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008113
CAS Number: 100-82-3
EINECS: 202-891-3
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty