Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-nitrotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

B23542-14 B23542-14 B23542-36 B23542-22 B23542-36 B23542-22
AAAB23542-22EA 77.29 USD InStock
AAAB23542-22 CAAAAB23542-14 CAAAAB23542-36 AAAB23542-14 AAAB23542-36 CAAAAB23542-22
2-Chloro-4-nitrotoluene 98%
2-Chloro-4-nitrotoluene


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 260 °C
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00007210
CAS Number: 121-86-8
EINECS: 204-501-7
UN: 3457
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty