Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Sulfobenzoic acid monosodium salt 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 3-sulfobenzoateSodium 3-sulphobenzoate

B23556-14 B23556-14 B23556-22 B23556-36 B23556-36 B23556-22
AAAB23556-14EA 28.59 USD InStock
CAAAAB23556-22 AAAB23556-14 AAAB23556-36 AAAB23556-22 CAAAAB23556-14 CAAAAB23556-36
3-Sulfobenzoic acid monosodium salt 97%
3-Sulphobenzoic acid monosodium salt


Formula: C₇H₅NaO₅S
MW: 224.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066378
CAS Number: 17625-03-5
EINECS: 241-602-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty