Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrimethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trimethoxy(methyl)silane

B23594-AK B23594-AU B23594-AU B23594-AK
AAAB23594-AKEA 44.39 USD InStock
AAAB23594-AK AAAB23594-AU CAAAAB23594-AK CAAAAB23594-AU
Methyltrimethoxysilane 97%
Methyltrimethoxysilane


Formula: C₄H₁₂O₃Si
Boiling Pt: 102…103 °C
Melting Pt: 11 °C
Density: 0.955
Flash Pt: 9 °C(48 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008342
CAS Number: 1185-55-3
EINECS: 214-685-0
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty