Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-(-)-BINAP(S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

B23872-03 B23872-01 B23872-01 B23872-03 B23872-06 B23872-06
AAAB23872-01EA 49.78 USD InStock
AAAB23872-01 CAAAAB23872-03 AAAB23872-03 CAAAAB23872-01 CAAAAB23872-06 AAAB23872-06
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl


Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…241 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-56-5
Merck Index: 13,01223
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty