Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-o-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Fluoro-2-anisaldehyde5-Fluoro-2-methoxybenzaldehyde

B24684-06 B24684-14 B24684-03 B24684-14 B24684-03 B24684-06
AAAB24684-14EA 489.99 USD InStock
CAAAAB24684-14 CAAAAB24684-03 AAAB24684-14 AAAB24684-03 CAAAAB24684-06 AAAB24684-06
5-Fluoro-o-anisaldehyde 98%
5-Fluoro-o-anisaldehyde


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143458
CAS Number: 19415-51-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty