Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methylcyclopentanol 98%

Supplier: Thermo Scientific Chemicals
Warning

B24744-22 B24744-06 B24744-22 B24744-14 B24744-14 B24744-06
AAAB24744-14EA 283.8 USD InStock
CAAAAB24744-22 CAAAAB24744-06 AAAB24744-14 AAAB24744-06 AAAB24744-22 CAAAAB24744-14
1-Methylcyclopentanol 98%
1-Methylcyclopentanol


Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 135…136 °C
Melting Pt: 36…37 °C
Density: 0.904
Flash Pt: 40 °C(105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001364
CAS Number: 1462-03-9
EINECS: 215-963-4
UN: 1987
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty