Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Dibenzoyl-D-tartaric acid, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2S,3S)-(+)-Di-O-benzoyltartaric acid

B24754-09 B24754-18 B24754-09 B24754-30 B24754-18 B24754-30
AAAB24754-09EA 27.29 USD InStock
AAAB24754-09 AAAB24754-18 CAAAAB24754-09 AAAB24754-30 CAAAAB24754-30 CAAAAB24754-18
(+)-Dibenzoyl-D-tartaric acid, anhydrous 99%
(+)-Dibenzoyl-D-tartaric acid
For chiral resolution.


Formula: C₁₈H₁₄O₈
MW: 358.31
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00063222
CAS Number: 17026-42-5
EINECS: 241-097-1
Order Now
Specification Test Results
Optical rotation +120° (c=5 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty