Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Adamantylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

B24770-03 B24770-06 B24770-06 B24770-03
AAAB24770-06EA 439.99 USD InStock
CAAAAB24770-06 AAAB24770-06 AAAB24770-03 CAAAAB24770-03
1-Adamantylacetonitrile 97%
1-Adamantylacetonitrile


Formula: C₁₂H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD00194034
CAS Number: 16269-13-9
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty