Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Pyridyloxyacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B24780-14 B24780-14 B24780-03 B24780-06 B24780-06 B24780-03
AAAB24780-06EA 288.99 USD InStock
CAAAAB24780-06 AAAB24780-06 CAAAAB24780-14 CAAAAB24780-03 AAAB24780-14 AAAB24780-03
3-Pyridyloxyacetic acid 98%
3-Pyridyloxyacetic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD04039772
CAS Number: 86649-57-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty