Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Glutamic acid 5-methyl ester 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Glu(OMe)-OH

B24859-14 B24859-14 B24859-22 B24859-06 B24859-06 B24859-22
AAAB24859-06EA 36.49 USD InStock
AAAB24859-06 CAAAAB24859-06 CAAAAB24859-22 AAAB24859-14 AAAB24859-22 CAAAAB24859-14
L-Glutamic acid 5-methyl ester 99%
L-Glutamic acid 5-methyl ester


Formula: C₆H₁₁NO₄
MW: 161.2 g/mol
Melting Pt: 182 °C (dec.)
Storage Temperature: Refrigerator
EINECS: 216-110-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty