Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Naphthalenethiol 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Thionaphthol

B24876-03 B24876-14 B24876-03 B24876-06 B24876-14 B24876-06
AAAB24876-03EA 62.78 USD InStock
CAAAAB24876-06 AAAB24876-03 AAAB24876-14 AAAB24876-06 CAAAAB24876-03 CAAAAB24876-14
1-Naphthalenethiol 99%
1-Naphthalenethiol


Formula: C₁₀H₈S
Storage Temperature: Ambient
MDL Number: MFCD00039599
CAS Number: 529-36-2
EINECS: 208-462-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty